missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl Phthalyl Ethyl Glycolate, TRC

Artikelnummer. 30135317
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135317 2.5 g 2.5g
30133615 250 mg -
17991175 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30135317 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9373552.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 84-72-0
Molekylær formel C14H16O6
Molekylvægt (g/mol) 280.27
Synonym Phthalic Acid, Ethyl Ester-(ethyl glycolate), 1,2-Benzenedicarboxylic acid, 1-(2-ethoxy-2-oxoethyl) 2-ethyl ester (ACI), 1,2-Benzenedicarboxylic acid, 2-ethoxy-2-oxoethyl ethyl ester (9CI), Glycolic acid, ethyl ester, ethyl phthalate (8CI), Phthalic acid, ethyl ester, ester with ethyl glycolate (6CI, 7CI, 8CI), 2-O-(2-Ethoxy-2-oxoethyl) 1-O-ethyl benzene-1,2-dicarboxylate, Carbethoxymethyl ethyl phthalate, Diethyl o-carboxybenzoyloxyacetate, Ethoxycarbonylmethyl ethyl phthalate, Ethyl 2-ethoxy-2-oxoethyl phthalate, Ethyl carbethoxymethyl phthalate, Ethyl ethoxycarbonylmethyl phthalate, Ethyl phthalyl ethyl glycolate, Santicizer E-15, Phthalic Acid, Ethyl Ester-(ethyl glycolate), 1,2-Benzenedicarboxylic acid, 1-(2-ethoxy-2-oxoethyl) 2-ethyl ester (ACI), 1,2-Benzenedicarboxylic acid, 2-ethoxy-2-oxoethyl ethyl ester (9CI), Glycolic acid, ethyl ester, ethyl phthalate (8CI), Phthalic acid, ethyl ester, ester with ethyl glycolate (6CI, 7CI, 8CI), 2-O-(2-Ethoxy-2-oxoethyl) 1-O-ethyl benzene-1,2-dicarboxylate, Carbethoxymethyl ethyl phthalate, Diethyl o-carboxybenzoyloxyacetate, Ethoxycarbonylmethyl ethyl phthalate, Ethyl 2-ethoxy-2-oxoethyl phthalate, Ethyl carbethoxymethyl phthalate, Ethyl ethoxycarbonylmethyl phthalate, Santicizer E-15
IUPAC navn 2-O-(2-ethoxy-2-oxoethyl) 1-O-ethyl benzene-1,2-dicarboxylate
SMIL CCOC(=O)COC(=O)c1ccccc1C(=O)OCC

Tekniske data

Kemisk navn eller materiale Ethyl Phthalyl Ethyl Glycolate
Mængde 2.5 g
InChI formel InChI=1S/C14H16O6/c1-3-18-12(15)9-20-14(17)11-8-6-5-7-10(11)13(16)19-4-2/h5-8H,3-4,9H2,1-2H3
Formel vægt 280.0947
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.