Learn More
Ethyl Palmitate, Sigma-Aldrich™
Product Code. 15870611
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250g
Product Code. Quantity Unit Size
15870611 250 mL 250g
1 options
Product Code. 15870611 Supplier Sigma Aldrich Supplier No. 8187750250
 Restricted Item
Forventet forsendelse: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 628-97-7
Specifications
Melting Point 23°C to 26°C
Density 0.86 g/cm3 at 20°C
Boiling Point 192°C to 194°C/13 hPa
Molecular Formula CH3(CH2)14COOC2H5
MDL Number MFCD00008996
Quantity 250 mL
Synonym Ethyl palmitate, Palmitic acid ethyl ester, Ethyl palmitate
Molecular Weight (g/mol) 284.48
Grade Research