Learn More
Ethyl nitroacetate, 97%, Thermo Scientific Chemicals
Catalog Number 11468930
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
626-35-7
C4H7NO4
133.103
MFCD00007403
FTKASJMIPSSXBP-UHFFFAOYSA-N
ethyl nitroacetate, nitroacetic acid ethyl ester, acetic acid, nitro-, ethyl ester, nitro ethyl acetate, ethylnitroacetate, acetic acid, 2-nitro-, ethyl ester, ethylnitroacetat, ethyinitroacetate, ethyl-2-nitroacetate, zlchem 269
69379
ethyl 2-nitroacetate
CCOC(=O)C[N+](=O)[O-]
626-35-7
C4H7NO4
133.103
MFCD00007403
FTKASJMIPSSXBP-UHFFFAOYSA-N
ethyl nitroacetate, nitroacetic acid ethyl ester, acetic acid, nitro-, ethyl ester, nitro ethyl acetate, ethylnitroacetate, acetic acid, 2-nitro-, ethyl ester, ethylnitroacetat, ethyinitroacetate, ethyl-2-nitroacetate, zlchem 269
69379
ethyl 2-nitroacetate
CCOC(=O)C[N+](=O)[O-]
Ethyl nitroacetate has been used in the synthesis of γ-oxoacids via Michael addition reaction with α,β-unsaturated ketones, fuctionalization of C4-position on pyrimidine and C6-position on 2-deoxyguanosine to produce novel nucleosides, facile synthesis of α,α-diisobutylglycine, synthesis of DL-4,4-difluoroglutamic acid. It is also used as an intermediate in the preparation of unsubstituted amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl nitroacetate has been used in the synthesis of γ-oxoacids via Michael addition reaction with α,β-unsaturated ketones, fuctionalization of C4-position on pyrimidine and C6-position on 2′-deoxyguanosine to produce novel nucleosides, facile synthesis of α,α-diisobutylglycine, synthesis of DL-4,4-difluoroglutamic acid. It is also used as an intermediate in the preparation of unsubstituted amino acids.
Solubility
Slightly soluble in water. Soluble in chloroform and ethyl acetate.
Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Density 1.202
Boiling Point 93°C to 95°C (10 mmHg)
Flash Point 92°C (197°F)
Odor Weak
Linear Formula O2NCH2CO2CH2CH3
Refractive Index 1.424
Quantity 1 g
Beilstein 1210027
Solubility Information Slightly soluble in water. Soluble in chloroform and ethyl acetate.
Formula Weight 133.11
Percent Purity 97%
Chemical Name or Material Ethyl nitroacetate