missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl Nicotinate-d4, TRC

Artikelnummer. 30374268
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30374268 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374268 Leverandør Toronto Research Chemicals Leverandørnr. TRCE925128500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl Nicotinate-d4
CAS 66148-16-1
Molekylær formel C8 2H4 H5 N O2
Mængde 500 mg
Synonym 3-Pyridine-2,4,5,6-d4-carboxylic acid, ethyl ester, 3-Pyridine-2,4,5,6-d4-carboxylic acid ethyl ester, Tetradeuterionicotinic acid ethyl ester, Ethyl 3-pyridinecarboxylate-2,4,5,6-d4, Ethyl nicotinate-d4, Ethyl nicotinate-2,4,5,6-D4
InChI formel InChI=1S/C8H9NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h3-6H,2H2,1H3/i3D,4D,5D,6D
SMIL [2H]c1nc([2H])c(C(=O)OCC)c([2H])c1[2H]
IUPAC navn ethyl 2,4,5,6-tetradeuteriopyridine-3-carboxylate
Molekylvægt (g/mol) 155.19
Formel vægt 155.0884
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.