missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl Linoleate-d5, TRC

Artikelnummer. 30383154
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30383154 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30383154 Leverandør Toronto Research Chemicals Leverandørnr. TRCE924302250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl Linoleate-d5
CAS 1331665-14-5
Molekylær formel C20H31D5O2
Mængde 250 mg
Synonym (9Z,12Z)-9,12-Octadecadienoic Acid Ethyl Ester-d5, (Z,Z)-9,12-Octadecadienoic Acid Ethyl Ester-d5, Linoleic Acid Ethyl Ester-d5, Ethyl (Z,Z)-9,12-Octadecadienoate-d5, Ethyl cis,cis-9,12-Octadecadienoate-d5, Ethyl Linolate-d5, Mandenol-d5, Nikkol VF-E-d5
InChI formel InChI=1S/C20H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h8-9,11-12H,3-7,10,13-19H2,1-2H3/b9-8-,12-11-/i2D3,4D2
SMIL O=C(OC([2H])([2H])C([2H])([2H])[2H])CCCCCCC/C=C\C/C=C\CCCCC
Molekylvægt (g/mol) 313.53
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.