Få mere at vide
Ethyl Isobutyrylacetate-d6, TRC
Artikelnummer. 30369610
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30369610 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369610 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92130250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Isobutyrylacetate-d6
CAS 1216475-83-0
Molekylær formel C8H8D6O3
Mængde 50 mg
Synonym Ethyl 5,5,5-trideuterio-3-oxo-4-(trideuteriomethyl)pentanoate, Ethyl isobutyrylacetate-d6, 3-Oxo-4-(methyl-d3)pentanoic-5,5,5-d3 acid ethyl ester, Ethyl 4-(methyl-d3)-3-oxopentanoate-5,5,5-d3, Ethyl 4-methyl-3-oxovalerate-d6, Ethyl 4-methyl-3-oxopentanoate-D6 (di(methyl-D3)), 4-(Methyl-d3)-3-oxo-5,5,5-d3-valeric acid ethyl ester
InChI formel InChI=1S/C8H14O3/c1-4-11-8(10)5-7(9)6(2)3/h6H,4-5H2,1-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])C(C(=O)CC(=O)OCC)C([2H])([2H])[2H]
IUPAC navn ethyl 5,5,5-trideuterio-3-oxo-4-(trideuteriomethyl)pentanoate
Molekylvægt (g/mol) 164.23
Formel vægt 164.132