missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl Isobutyrylacetate-d6, TRC

Artikelnummer. 30376998
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376998 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376998 Leverandør Toronto Research Chemicals Leverandørnr. TRCE921302500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl Isobutyrylacetate-d6
CAS 1216475-83-0
Molekylær formel C8H8D6O3
Mængde 500 mg
Synonym Ethyl 5,5,5-trideuterio-3-oxo-4-(trideuteriomethyl)pentanoate, Ethyl isobutyrylacetate-d6, 3-Oxo-4-(methyl-d3)pentanoic-5,5,5-d3 acid ethyl ester, Ethyl 4-(methyl-d3)-3-oxopentanoate-5,5,5-d3, Ethyl 4-methyl-3-oxovalerate-d6, Ethyl 4-methyl-3-oxopentanoate-D6 (di(methyl-D3)), 4-(Methyl-d3)-3-oxo-5,5,5-d3-valeric acid ethyl ester
InChI formel InChI=1S/C8H14O3/c1-4-11-8(10)5-7(9)6(2)3/h6H,4-5H2,1-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])C(C(=O)CC(=O)OCC)C([2H])([2H])[2H]
IUPAC navn ethyl 5,5,5-trideuterio-3-oxo-4-(trideuteriomethyl)pentanoate
Molekylvægt (g/mol) 164.23
Formel vægt 164.132
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Sekretesspolicy.