Få mere at vide
Ethyl Isobutyrate, TRC
Artikelnummer. 30373791
Skift visning
Mængde:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30373791 25 g
30384707 100 g
30371198 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30373791 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92130525G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 97-62-1
Molekylær formel C6 H12 O2
Molekylvægt (g/mol) 116.16
Synonym Isobutyric acid, ethyl ester (6CI, 8CI), 2-Methylpropanoic acid ethyl ester, 2-Methylpropionic acid ethyl ester, Ethyl 2,2-dimethylacetate, Ethyl 2-methyl-1-propanoate, Ethyl 2-methylpropanoate, Ethyl 2-methylpropionate, Ethyl isobutanoate, Ethyl isobutyrate, NSC 97194
IUPAC navn ethyl 2-methylpropanoate
SMIL CCOC(=O)C(C)C
Tekniske data
Kemisk navn eller materiale Ethylisobutyrate
Mængde 25 g
InChI formel InChI=1S/C6H12O2/c1-4-8-6(7)5(2)3/h5H,4H2,1-3H3
Formel vægt 116.0837