Få mere at vide
Ethyl Hexanoate-d11, CDN
Artikelnummer. 30313439
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313439 0.25 g
30310094 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313439 Leverandør CDN Leverandørnr. CDND70600.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Caproate-d11
CAS 2159-19-5
Molekylær formel C8 2H11 H5 O2
Mængde 0.25 g
InChI formel InChI=1 S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3/i1D3,3D2,5D2,6D2,7D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OCC
IUPAC navn ethyl 2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexanoate
Molekylvægt (g/mol) 155.28
Formel vægt 155.1841 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity