missing translation for 'onlineSavingsMsg'
Learn More

Ethyl fluoroacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-130252
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
459-72-3
C4H7FO2
106.1
MFCD00000450
VCYZVXRKYPKDQB-UHFFFAOYSA-N
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
9988
ethyl 2-fluoroacetate
CCOC(=O)CF
This item is not returnable. View return policy
459-72-3
C4H7FO2
106.1
MFCD00000450
VCYZVXRKYPKDQB-UHFFFAOYSA-N
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
9988
ethyl 2-fluoroacetate
CCOC(=O)CF

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.0900g/mL
Boiling Point 119.2°C (753.0 mmHg)
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula FCH2CO2C2H5
Refractive Index 1.3745 to 1.3765
Quantity 5 g
Beilstein 02, 193
Fieser 05,304
Merck Index 15, 4199
Specific Gravity 1.09
Solubility Information Solubility in water: decomposes.
Formula Weight 106.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl fluoroacetate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl fluoroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'