Få mere at vide
Ethyl Diphenylacetate, Mikromol™
Artikelnummer. 30050718
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050718 100 mg
30051140 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050718 Leverandør Mikromol Leverandørnr. MM0025.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl Diphenylacetate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Diphenylacetate
CAS 3468-99-3
Analyt- eller komponentnavne Ethyl Diphenylacetate
Forsendelsestilstand Room Temperature
Molekylær formel C16 H16 O2
Certificeringer/compliance ISO 17025
Synonym Ethyl Diphenylacetate
InChI formel InChI=1S/C16H16O2/c1-2-18-16(17)15(13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12,15H,2H2,1H3
SMIL CCOC(=O)C(c1ccccc1)c2ccccc2
IUPAC navn ethyl 2,2-diphenylacetate
Molekylvægt (g/mol) 240.3
Mængde 100 mg
Formel vægt 240.115 g/mol