missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dimethylpyruvate, 97%, Thermo Scientific Chemicals

Catalog Number p-7035689
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
20201-24-5
C7H12O3
144.17
MFCD00009122
CKTYYUQUWFEUCO-UHFFFAOYSA-N
ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate
88406
ethyl 3-methyl-2-oxobutanoate
CCOC(=O)C(=O)C(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
20201-24-5
C7H12O3
144.17
MFCD00009122
CKTYYUQUWFEUCO-UHFFFAOYSA-N
ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate
88406
ethyl 3-methyl-2-oxobutanoate
CCOC(=O)C(=O)C(C)C

Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.989
Boiling Point 62°C (11 mmHg)
Flash Point 43°C (109°F)
Refractive Index 1.41
Quantity 5 g
UN Number UN3272
Beilstein 1756668
Solubility Information Slightly soluble in water.
Formula Weight 144.17
Percent Purity 97%
Chemical Name or Material Ethyl dimethylpyruvate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'