missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl (Diethoxyphosphoryl)formate, Mikromol™

Artikelnummer. 30049281
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049281 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30049281 Leverandør Mikromol Leverandørnr. MM3500.02

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl (Diethoxyphosphoryl)formate
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl diethoxyphosphinylformate
CAS 1474-78-8
Analyt- eller komponentnavne Ethyl diethoxyphosphinylformate
Forsendelsestilstand Room Temperature
Molekylær formel C7 H15 O5 P
Certificeringer/compliance ISO 9001
Synonym Foscarnet Sodium Hexahydrate Imp. D (EP), Foscarnet Imp. D (EP), Foscarnet USP Related Compound D, Foscarnet USP RC D, Phosphinecarboxylic acid, 1,1-diethoxy-, ethyl ester, 1-oxide, Formic acid, phosphono-, triethyl ester (7CI,8CI), Phosphinecarboxylic acid, diethoxy-, ethyl ester, oxide (9CI), Diethyl (ethoxycarbonyl)phosphonate, Diethyl carbethoxyphosphonate, Ethyl diethylphosphonoformate, NSC 108684, Triethyl carboxyphosphonate, Triethyl phosphonoformate, Ethyl (Diethoxyphosphoryl)formate, O,O-Diethyl ethoxycarbonylphosphonate, Foscarnet Related Compound D, Foscarnet Sodium Hexahydrate Impurity D, Foscarnet Impurity D
InChI formel InChI=1S/C7H15O5P/c1-4-10-7(8)13(9,11-5-2)12-6-3/h4-6H2,1-3H3
SMIL CCOC(=O)P(=O)(OCC)OCC
IUPAC navn ethyl diethoxyphosphorylformate
Molekylvægt (g/mol) 210.16
Mængde 100 mg
Formel vægt 210.0657 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.