Få mere at vide
Ethyl-d5 Vanillin, TRC
Artikelnummer. 30378189
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30378189 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378189 Leverandør Toronto Research Chemicals Leverandørnr. TRCE932102100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl-d5 Vanillin
CAS 1335401-74-5
Renhedsgrad noter HPLC
Molekylær formel C9 2H5 H5 O3
Mængde 100 mg
Synonym Benzaldehyde, 3-(ethoxy-1,1,2,2,2-d5)-4-hydroxy-, Ethyl-d5 vanillin, 3-Ethoxy-4-hydroxybenzaldehyde-d5, 3-(Ethoxy-d5)-4-hydroxybenzaldehyde, 3-Ethoxy-4-hydroxybenzaldehyde-D5 (ethoxy-D5), 3-(Ethyl-d5)vanillin, 4-Hydroxy-3-(ethoxy-d5)benzaldehyde, Arovanillon-d5, Bourbonal-d5, Ethylvanillin, -d5
InChI formel InChI=1S/C9H10O3/c1-2-12-9-5-7(6-10)3-4-8(9)11/h3-6,11H,2H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1cc(C=O)ccc1O
IUPAC navn 4-hydroxy-3-(1,1,2,2,2-pentadeuterioethoxy)benzaldehyde
Molekylvægt (g/mol) 171.2
Formel vægt 171.0944
Procent renhed >95