Få mere at vide
Ethyl-d5 beta-D-glucuronide, TRC
Artikelnummer. 30377143
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30377143 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377143 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91842710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl beta-D-glucuronide-D5
CAS 1135070-98-2
Renhedsgrad noter HPLC
Molekylær formel C8 2H5 H9 O7
Mængde 10 mg
Synonym 6-Ethoxy-d5-3,4,5-trihydroxy-tetrahydropyran-2-carboxylic Acid, Ethyl-d5 beta-D-glucuronide
InChI formel InChI=1S/C8H14O7/c1-2-14-8-5(11)3(9)4(10)6(15-8)7(12)13/h3-6,8-11H,2H2,1H3,(H,12,13)/t3-,4-,5+,6-,8+/m0/s1/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O
IUPAC navn (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-(1,1,2,2,2-pentadeuterioethoxy)oxane-2-carboxylic acid
Molekylvægt (g/mol) 227.22
Formel vægt 227.1053
Procent renhed >95