Få mere at vide
Ethyl-d5-amine HCl, CDN
Artikelnummer. 30322409
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322409 1 g -
30312165 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322409 Leverandør CDN Leverandørnr. CDND00161G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl-D5-Amine Hydrochloride
CAS 284474-81-3
Molekylær formel C2 D5 H2 N . Cl H
Mængde 1 g
Synonym Ethylamine-d5 Hydrochloride, Ethyl-d5-amine HCl
InChI formel InChI=1 S/C2H7N.ClH/c1-2-3;/h2-3H2,1H3;1 H/i1D3,2D2;
SMIL Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])N
IUPAC navn 1,1,2,2,2-pentadeuterioethanamine;hydrochloride
Molekylvægt (g/mol) 86.58
Formel vægt 86.0659 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity