Få mere at vide
Ethyl-d5 Alcohol, CDN
Artikelnummer. 30311369
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311369 5 g
30321698 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311369 Leverandør CDN Leverandørnr. CDND01085G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethanol-d5
CAS 1859-08-1
Molekylær formel C2 2H5 H O
Mængde 5 g
InChI formel InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])O
IUPAC navn 1,1,2,2,2-pentadeuterioethanol
Molekylvægt (g/mol) 51.1
Formel vægt 51.0732 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity