Få mere at vide
Ethyl-d5 4-Aminobenzoate, CDN
Artikelnummer. 30322372
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322372 0.1 g 0.1g
30328460 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322372 Leverandør CDN Leverandørnr. CDND68640.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzocaine-D5 (ethyl-D5)
CAS 1219803-76-5
Molekylær formel C9 D5 H6 N O2
Mængde 0.1 g
Synonym Benzocaine-d5, Benzocaine-D5 (ethyl-D5), 4-Aminobenzoic acid ethyl-d5 ester, Ethyl-d5 4-aminobenzoate
InChI formel InChI=1 S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6 H,2,10H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])OC(=O)c1ccc(N)cc1
IUPAC navn 1,1,2,2,2-pentadeuterioethyl 4-aminobenzoate
Molekylvægt (g/mol) 170.22
Formel vægt 170.11 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity