Få mere at vide
Ethyl-d3 Chloroformate, TRC
Artikelnummer. 30353507
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30353507 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353507 Leverandør Toronto Research Chemicals Leverandørnr. TRCC215211100MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl-d3 Chloroformate
CAS 70907-86-7
Molekylær formel C3 D3 H2 Cl O2
Mængde 100 mg
Synonym Carbonochloridic Acid Ethyl-d3 Ester, Cathyl Chloride, Chloroformic Acid Ethyl-d3 Ester, Chloro Ethyl-d3 Formate, Chlorocarbonic Acid Ethyl-d3 Ester, Ethoxycarbonyl-d3 Chloride, Ethyl-d3 Carbonochloridate, Ethyl-d3 Chloridocarbonate, Ethyl-d3 Chlorocarbonate
InChI formel InChI=1S/C3H5ClO2/c1-2-6-3(4)5/h2H2,1H3/i1D3
SMIL [2H]C([2H])([2H])COC(=O)Cl
IUPAC navn 2,2,2-trideuterioethyl carbonochloridate
Molekylvægt (g/mol) 111.542
Formel vægt 111.017