Få mere at vide
Ethyl Cyclopropylcarboxylate-d5 (Major), TRC
Artikelnummer. 30374208
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30374208 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374208 Leverandør Toronto Research Chemicals Leverandørnr. TRCE90905325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Cyclopropylcarboxylate-d5 (Major)
CAS 1794783-51-9
Molekylær formel C6H5D5O2
Mængde 25 mg
Synonym Cyclopropanecarboxylic Acid Ethyl Ester-d5, Ethyl Cyclopropanecarboxylate-d5, NSC 60696-d5
InChI formel InChI=1S/C6H10O2/c1-2-8-6(7)5-3-4-5/h5H,2-4H2,1H3/i3D2,4D2,5D
SMIL [2H]C1([2H])C([2H])([2H])C1([2H])C(=O)OCC
IUPAC navn ethyl 1,2,2,3,3-pentadeuteriocyclopropane-1-carboxylate
Molekylvægt (g/mol) 119.17
Formel vægt 119.0995