Learn More
Ethyl cyclohexylideneacetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.
Solubility
Soluble in dichloromethane, ethyl acetate.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.
Solubility
Soluble in dichloromethane, ethyl acetate.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Boiling Point | 48°C to 49°C (0.02 mmHg) |
Quantity | 5 g |
Solubility Information | Soluble in dichloromethane,ethyl acetate. |
Formula Weight | 168.24 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl cyclohexylideneacetate |