Få mere at vide
Ethyl Cyanoglyxylate-2-oxyme Potassium Salt, TRC
Artikelnummer. 30371028
Skift visning
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371028 5 g
30370229 10 g
30369838 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30371028 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9378505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 158014-03-0
Molekylær formel C5H6N2O3K
Molekylvægt (g/mol) 181.2
Synonym Acetic acid, 2-cyano-2-(hydroxyimino)-, ethyl ester, potassium salt (1:1) (ACI), Acetic acid, cyano(hydroxyimino)-, ethyl ester, potassium salt (9CI), 2-Cyano-2-(hydroxyimino)acetic acid ethyl ester potassium salt, Ethyl 2-(hydroxyimino)cyanoacetate potassium salt, Ethyl 2-cyano-2-(hydroxyimino)acetate potassium salt, Ethyl 2-cyano-2-oximinoacetate potassium salt, Ethyl cyanoglyoxylate oxime potassium salt, Ethyl cyanoglyoxylate-2-oxime potassium salt, Ethyl α-cyano-α-oximinoacetate potassium salt
SMIL [K+].CCOC(=O)C(=N[O-])C#N
Tekniske data
Kemisk navn eller materiale Ethyl Cyanoglyxylate-2-oxyme Potassium Salt
Mængde 5 g
InChI formel InChI=1S/C5H6N2O3.K/c1-2-10-5(8)4(3-6)7-9;/h9H,2H2,1H3;/q;+1/p-1/b7-4+;
Formel vægt 179.9937