Få mere at vide
Ethyl Clofibrate-d4 (4-chlorophenyl-d4), CDN
Artikelnummer. 30318947
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318947 0.01 g
30317866 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318947 Leverandør CDN Leverandørnr. CDND64150.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clofibrate-d4
CAS 1189654-03-2
Molekylær formel C12 2H4 H11 Cl O3
Mængde 0.01 g
InChI formel InChI=1 S/C12H15ClO3/c1-4-15-11(14)12(2,3)16-10-7-5-9(13)6-8-10/h5-8 H,4H2,1-3H3/i5D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c(OC(C)(C)C(=O)OCC)c([2 H])c([2 H])c1Cl
IUPAC navn ethyl 2-(4-chloro-2,3,5,6-tetradeuteriophenoxy)-2-methylpropanoate
Molekylvægt (g/mol) 246.72
Formel vægt 246.0961 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity