missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl Caproate-d11, TRC

Artikelnummer. 30376367
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376367 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376367 Leverandør Toronto Research Chemicals Leverandørnr. TRCE90160250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl Caproate-d11
CAS 2159-19-5
Molekylær formel C8 2H11 H5 O2
Mængde 50 mg
Synonym Hexanoic-d11 acid, ethyl ester (7CI, 8CI, ACI), Ethyl hexanoate-2,2,3,3,4,4,5,5,6,6,6-d11, Hexanoic-2,2,3,3,4,4,5,5,6,6,6-d11 acid ethyl ester, Hexanoic-d11 acid ethyl ester, Ethyl hexanoate-d11, Hexanoic Acid Ethyl Ester-D11 (hexanoic-D11), Ethyl Caproate-d11
InChI formel InChI=1S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3/i1D3,3D2,5D2,6D2,7D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OCC
IUPAC navn ethyl 2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexanoate
Molekylvægt (g/mol) 155.28
Formel vægt 155.1841
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.