Få mere at vide
Ethyl Caproate-d11, TRC
Artikelnummer. 30372849
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30372849 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372849 Leverandør Toronto Research Chemicals Leverandørnr. TRCE90160225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Caproate-d11
CAS 2159-19-5
Molekylær formel C8 2H11 H5 O2
Mængde 25 mg
Synonym Hexanoic-d11 acid, ethyl ester (7CI, 8CI, ACI), Ethyl hexanoate-2,2,3,3,4,4,5,5,6,6,6-d11, Hexanoic-2,2,3,3,4,4,5,5,6,6,6-d11 acid ethyl ester, Hexanoic-d11 acid ethyl ester, Ethyl hexanoate-d11, Hexanoic Acid Ethyl Ester-D11 (hexanoic-D11), Ethyl Caproate-d11
InChI formel InChI=1S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3/i1D3,3D2,5D2,6D2,7D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OCC
IUPAC navn ethyl 2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexanoate
Molekylvægt (g/mol) 155.28
Formel vægt 155.1841