missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl Butyrate-d3, TRC

Artikelnummer. 30370707
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30370707 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370707 Leverandør Toronto Research Chemicals Leverandørnr. TRCE90069250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl Butyrate-d3
CAS 113435-99-7
Molekylær formel C6 2H3 H9 O2
Mængde 50 mg
Synonym Butanoic-4,4,4-d3 acid, ethyl ester (9CI), Butanoic-4,4,4-d3 acid ethyl ester, Butyric Acid-4,4,4-d3 Ethyl Ester, Ethyl Butyrate-d3, Ethyl Butyrate-4,4,4-d3, Ethyl Butanoate-4,4,4-d3, Butyric Acid Ethyl Ester-D3 (butyric-4,4,4-D3)
InChI formel InChI=1S/C6H12O2/c1-3-5-6(7)8-4-2/h3-5H2,1-2H3/i1D3
SMIL [2H]C([2H])([2H])CCC(=O)OCC
IUPAC navn ethyl 4,4,4-trideuteriobutanoate
Molekylvægt (g/mol) 119.18
Formel vægt 119.1026
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.