Få mere at vide
Ethyl Biscoumacetate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl Biscoumacetate |
| CAS | 548-00-5 |
| Analyt- eller komponentnavne | Ethyl Biscoumacetate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H16 O8 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Acetic acid, bis(4-hydroxy-2-oxo-2H-1-benzopyran-3-yl)-, ethyl ester (6CI,7CI,8CI), 3,3'-(Carboxymethylene)bis(4-hydroxycoumarin) ethyl ester, BOEA, Bis(4-hydroxy-2-oxo-2H-1-benzopyran-3-yl)acetic acid ethyl ester, Bis(4-hydroxy-3-coumarin)acetic acid ethyl ester, Dicumacyl, Ethyl 4,4'-dihydroxydicoumarinyl-3,3'-acetate, Ethyl bis(4-hydroxycoumarinyl)acetate, Ethyl biscoumacetate, NSC 36366, Neodicoumarin, Neodicoumarol, Neodicumarinum, Neodicumarol, Pelentan, Stabilene, Trombarin, Trombil, Trombolysan, Tromexan, Tromexan ethyl acetate, 2H-1-Benzopyran-3-acetic acid, 4-hydroxy-alpha-(4-hydroxy-2-oxo-2H-1-benzopyran-3-yl)-2-oxo-, ethyl ester |
| InChI formel | InChI=1S/C22H16O8/c1-2-28-20(25)15(16-18(23)11-7-3-5-9-13(11)29-21(16)26)17-19(24)12-8-4-6-10-14(12)30-22(17)27/h3-10,15,23-24H,2H2,1H3 |
| Løsningstype | Neat |
| SMIL | CCOC(=O)C(C1=C(O)c2ccccc2OC1=O)C3=C(O)c4ccccc4OC3=O |
| Vis mere |