missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzoylformate, 95%, Thermo Scientific Chemicals

Catalog Number p-3212685
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1603-79-8
C10H10O3
178.19
MFCD00009120
QKLCQKPAECHXCQ-UHFFFAOYSA-N
ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester
15349
CHEBI:84260
ethyl 2-oxo-2-phenylacetate
CCOC(=O)C(=O)C1=CC=CC=C1
This item is not returnable. View return policy
1603-79-8
C10H10O3
178.19
MFCD00009120
QKLCQKPAECHXCQ-UHFFFAOYSA-N
ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester
15349
CHEBI:84260
ethyl 2-oxo-2-phenylacetate
CCOC(=O)C(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Green
Density 1.1220g/mL
Boiling Point 138°C to 139°C (18.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COCO2CH2CH3
Refractive Index 1.5150 to 1.517
Quantity 5 g
Specific Gravity 1.122
Formula Weight 178.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl benzoylformate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'