Få mere at vide
Ethyl Benzenesulfonate, TRC
Artikelnummer. 30009498
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009498 100 mg
30005454 250 mg
30011338 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009498 Leverandør Toronto Research Chemicals Leverandørnr. TRCE708000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 515-46-8
Molekylær formel C8 H10 O3 S
Molekylvægt (g/mol) 186.23
Synonym Benzenesulfonic Acid Ethyl Ester
IUPAC navn ethyl benzenesulfonate
SMIL CCOS(=O)(=O)c1ccccc1
Tekniske data
Kemisk navn eller materiale Ethyl Benzenesulfonate
Mængde 100 mg
InChI formel InChI=1S/C8H10O3S/c1-2-11-12(9,10)8-6-4-3-5-7-8/h3-7H,2H2,1H3
Formel vægt 186.0351