Få mere at vide
Ethyl Arachidonate-d5, TRC
Artikelnummer. 30377604
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30377604 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377604 Leverandør Toronto Research Chemicals Leverandørnr. TRCE89940250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Arachidonate-d5
CAS 1331642-74-0
Molekylær formel C22 D5 H31 O2
Mængde 50 mg
Synonym (5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic Acid Ethyl Ester-d5, (all-Z)-5,8,11,14-Eicosatetraenoic Acid Ethyl Ester-d5, Arachidonic Acid Ethyl Ester-d5
InChI formel InChI=1S/C22H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24-4-2/h8-9,11-12,14-15,17-18H,3-7,10,13,16,19-21H2,1-2H3/b9-8-,12-11-,15-14-,18-17-/i2D3,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OC(=O)CCC\C=C/C\C=C/C\C=C/C\C=C/CCCCC
IUPAC navn 1,1,2,2,2-pentadeuterioethyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
Molekylvægt (g/mol) 337.55
Formel vægt 337.3029