Få mere at vide
Ethyl alpha-(p-Methoxyphenyl)-beta-(dimethylamino)propionate, TRC
Artikelnummer. 30004855
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004855 500 mg
30006531 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004855 Leverandør Toronto Research Chemicals Leverandørnr. TRCE925055500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 323176-93-8
Molekylær formel C14 H21 N O3
Molekylvægt (g/mol) 251.32
Synonym Benzeneacetic acid, α-[(dimethylamino)methyl]-4-methoxy-, ethyl ester, Ethyl α-(p-methoxyphenyl)-β-(dimethylamino)propionate, α-[(Dimethylamino)methyl]-4-methoxybenzeneacetic Acid Ethyl Ester, Venlafaxine EP Impurity B;
IUPAC navn ethyl 3-(dimethylamino)-2-(4-methoxyphenyl)propanoate
SMIL CCOC(=O)C(CN(C)C)c1ccc(OC)cc1
Tekniske data
Kemisk navn eller materiale Ethyl alpha-(p-Methoxyphenyl)-beta-(dimethylamino)propionate
Renhedsgrad noter HPLC
Mængde 500 mg
InChI formel InChI=1S/C14H21NO3/c1-5-18-14(16)13(10-15(2)3)11-6-8-12(17-4)9-7-11/h6-9,13H,5,10H2,1-4H3
Formel vægt 251.1521
Procent renhed >95%