Få mere at vide
Ethyl Acetoacetate-2,2,4,4,4-d5, CDN
Artikelnummer. 30320415
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320415 0.25 g -
30313680 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320415 Leverandør CDN Leverandørnr. CDND82020.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Acetoacetate-2,2,4,4,4-D5
CAS 55514-60-8
Molekylær formel C6 D5 H5 O3
Mængde 0.25 g
Synonym Butanoic-2,2,4,4,4-d5 acid, 3-oxo-, ethyl ester (9 CI), Ethyl 3-oxobutanoate-2,2,4,4,4-d5, Ethyl acetoacetate-d5, Ethyl Acetoacetate-2,2,4,4,4-D5, 1-Ethoxybutane-1,3-dione-2,2,4,4,4-d5, Acetoacetic-2,2,4,4,4-d5 acid ethyl ester, Acetoacetic acid ethyl ester-d5
InChI formel InChI=1 S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3/i2D3,4D2
SMIL [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])C(=O)OCC
IUPAC navn ethyl 2,2,4,4,4-pentadeuterio-3-oxobutanoate
Molekylvægt (g/mol) 135.1726
Formel vægt 135.0944 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity