Få mere at vide
Ethyl Acetate-d8, TRC
Artikelnummer. 30371056
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30371056 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371056 Leverandør Toronto Research Chemicals Leverandørnr. TRCE678351100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Acetate-d8
CAS 117121-81-0
Renhedsgrad noter GC
Molekylær formel C4 D8 O2
Mængde 100 mg
Synonym Ethyl-d5 Acetate-d3, Ethyl-1,1,2,2,2-d5 Ester Acetic-2,2,2-d3 Acid, Ethyl-d5 Ester Acetic-d3 Acid
InChI formel InChI=1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3/i1D3,2D3,3D2
SMIL [2H]C([2H])([2H])C(=O)OC([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,2,2,2-pentadeuterioethyl 2,2,2-trideuterioacetate
Molekylvægt (g/mol) 96.15
Formel vægt 96.1026
Procent renhed >95