Få mere at vide
Ethyl Acetate-d8, CDN
Artikelnummer. 30321208
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321208 1 g -
30310209 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321208 Leverandør CDN Leverandørnr. CDND54851G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl Acetate-d8
CAS 117121-81-0
Molekylær formel C4 D8 O2
Mængde 1 g
Synonym Ethyl-d5 Acetate-d3, Ethyl-1,1,2,2,2-d5 Ester Acetic-2,2,2-d3 Acid, Ethyl-d5 Ester Acetic-d3 Acid
InChI formel InChI=1 S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3/i1D3,2D3,3D2
SMIL [2 H]C([2 H])([2 H])C(=O)OC([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,2,2,2-pentadeuterioethyl 2,2,2-trideuterioacetate
Molekylvægt (g/mol) 96.15
Formel vægt 96.1026 g/mol
Procent renhed 99.5 atom % D, min 98% Chemical Purity