missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, ACS, ≥99.5%, Honeywell Riedel-de Haën™

Catalog Number p-7120415
Click to view available options
Quantity:
1 L
4 L
Packaging:
Aluminum Bottle
This item is not returnable. View return policy
141-78-6
C4H8O2
88.106
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
This item is not returnable. View return policy
141-78-6
C4H8O2
88.106
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -84°C
Color Colorless
Linear Formula CH3COOC2H5
UN Number UN1173
Beilstein 506104
Solubility Information 80g/L in water at 20°C
Formula Weight 88.11g/mol
Percent Purity ≥99.5%
Density 0.902g/mL at 25°C
Boiling Point 77°C
Quantity 1 L
Explosion Limit 2.2 to 11.5%
Ignition Point 801°C
Packaging Aluminum Bottle
Physical Form Liquid
Vapor Pressure 100mmHg (20°C)
Chemical Name or Material Ethyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethyl acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'