Få mere at vide
Ethyl Acetate-1,2-C2, SPEX CertiPrep™
Artikelnummer. 12989234 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
12989234 1 mL 1ml
1 options
Artikelnummer. 12989234 Leverandør SPEX Certiprep Leverandørnr. S1921
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Ethyl acetate-1,2,-13C284508-45-21000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale Ethyl acetate-1,2,-13C2
Emballage 2mL amber prescored glass ampule
CAS 84508-45-2, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C4H8O2
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle XEKOWRVHYACXOJ-NDLBAUGKSA-N
SMIL CCOC(=O)C
IUPAC navn ethyl acetate
Opløsningsmiddel eller Matrix Methanol-P&T
Molekylvægt (g/mol) 90.091
PubChem CID 12206864
Mængde 1 mL
Vis mere Vis mindre
Product intended for laboratory use only