Få mere at vide
Ethyl 8-Ethyl-5-oxo-2-(piperazin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate (Pipemidic Acid Ethyl Ester), Mikromol™
Beskrivelse
Ethyl 8-Ethyl-5-oxo-2-(piperazin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate (Pipemidic Acid Ethyl Ester)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 8-Ethyl-5-oxo-2-(piperazin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate (Pipemidic Acid Ethyl Ester) |
| CAS | 51940-43-3 |
| Analyt- eller komponentnavne | Ethyl 8-Ethyl-5-oxo-2-(piperazin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate (Pipemidic Acid Ethyl Ester) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H21 N5 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Ethyl 8-Ethyl-5-oxo-2-(piperazin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate (Pipemidic Acid Ethyl Ester), Pipemidic Acid Trihydrate Imp. E (EP), Pipemidic Acid Imp. E (EP), 8-Ethyl-5,8-dihydro-5-oxo-2-(1-piperazinyl)pyrido[2,3-d]pyrimidine-6-carboxylic acid ethyl ester, Ethyl pipemidate, Pipemidic Acid Trihydrate Impurity E, Pipemidic Acid Impurity E |
| InChI formel | InChI=1S/C16H21N5O3/c1-3-20-10-12(15(23)24-4-2)13(22)11-9-18-16(19-14(11)20)21-7-5-17-6-8-21/h9-10,17H,3-8H2,1-2H3 |
| SMIL | CCOC(=O)C1=CN(CC)c2nc(ncc2C1=O)N3CCNCC3 |
| IUPAC navn | ethyl 8-ethyl-5-oxo-2-piperazin-1-ylpyrido[2,3-d]pyrimidine-6-carboxylate |
| Vis mere |