Learn More
Ethyl 7-bromoheptanoate, 97%, BLD Pharm™
Tekniske data
Tekniske data
| CAS | 29823-18-5 |
| Molecular Formula | C9H17BrO2 |
| MDL Number | MFCD00037103 |
| Quantity | 10 g |
| SMILES | CCOC(=O)CCCCCCBr |
| Molecular Weight (g/mol) | 237.13 |
| Percent Purity | 97% |
| Chemical Name or Material | Ethyl 7-bromoheptanoate |