missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-chloro-5-cyano-2-(trifluoromethyl)nicotinate, 97%, Thermo Scientific™

Catalog Number p-7032261
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
175277-73-3
C10H6ClF3N2O2
278.615
MFCD00178758
PMDLSQHFIADYJD-UHFFFAOYSA-N
ethyl 6-chloro-5-cyano-2-trifluoromethyl nicotinate, ethyl 6-chloro-5-cyano-2-trifluoromethyl pyridine-3-carboxylate, ethyl 2-chloro-3-cyano-6-trifluoromethyl pyridine-5-carboxylate, 3-pyridinecarboxylicacid, 6-chloro-5-cyano-2-trifluoromethyl-, ethyl ester, ethyl 6-chloro-5-cyano-2-trifluoromethyl-nicotinate, ethyl 2-chloro-3-cyano-6-trifluoromethylpyridine-5-carboxylate, ethyl 6-chloranyl-5-cyano-2-trifluoromethyl pyridine-3-carboxylate, 6-chloro-5-cyano-2-trifluoromethyl-3-pyridinecarboxylic acid ethyl ester
2737159
ethyl 6-chloro-5-cyano-2-(trifluoromethyl)pyridine-3-carboxylate
CCOC(=O)C1=C(N=C(C(=C1)C#N)Cl)C(F)(F)F
This item is not returnable. View return policy
175277-73-3
C10H6ClF3N2O2
278.615
MFCD00178758
PMDLSQHFIADYJD-UHFFFAOYSA-N
ethyl 6-chloro-5-cyano-2-trifluoromethyl nicotinate, ethyl 6-chloro-5-cyano-2-trifluoromethyl pyridine-3-carboxylate, ethyl 2-chloro-3-cyano-6-trifluoromethyl pyridine-5-carboxylate, 3-pyridinecarboxylicacid, 6-chloro-5-cyano-2-trifluoromethyl-, ethyl ester, ethyl 6-chloro-5-cyano-2-trifluoromethyl-nicotinate, ethyl 2-chloro-3-cyano-6-trifluoromethylpyridine-5-carboxylate, ethyl 6-chloranyl-5-cyano-2-trifluoromethyl pyridine-3-carboxylate, 6-chloro-5-cyano-2-trifluoromethyl-3-pyridinecarboxylic acid ethyl ester
2737159
ethyl 6-chloro-5-cyano-2-(trifluoromethyl)pyridine-3-carboxylate
CCOC(=O)C1=C(N=C(C(=C1)C#N)Cl)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 78°C to 79°C
Quantity 250 mg
Formula Weight 278.62
Percent Purity 97%
Chemical Name or Material Ethyl 6-chloro-5-cyano-2-(trifluoromethyl)nicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'