missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl 5-Methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate, TRC

Artikelnummer. 30378127
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378127 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30378127 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92352510MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl 5-Methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
CAS 78756-03-3
Molekylær formel C15 H15 N3 O3
Mængde 10 mg
Synonym 4H-Imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid, 5,6-dihydro-5-methyl-6-oxo-, ethyl ester, Ethyl 5-Methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate, Ro 14-7437, Imp. E (EP)
InChI formel InChI=1S/C15H15N3O3/c1-3-21-15(20)13-12-8-17(2)14(19)10-6-4-5-7-11(10)18(12)9-16-13/h4-7,9H,3,8H2,1-2H3
SMIL CCOC(=O)c1ncn2c1CN(C)C(=O)c3ccccc23
IUPAC navn ethyl 5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
Molekylvægt (g/mol) 285.3
Formel vægt 285.1113
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.