Få mere at vide
Ethyl 5-Ethyl-8-oxo-5,8-dihydro-1,3-dioxolo[4,5-g]quinoline-7-carboxylate (Oxolinic Acid Ethyl Ester), Mikromol™
Beskrivelse
Ethyl 5-Ethyl-8-oxo-5,8-dihydro-1,3-dioxolo[4,5-g]quinoline-7-carboxylate (Oxolinic Acid Ethyl Ester)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Oxolinic acid ethyl ester |
| CAS | 16172-03-5 |
| Analyt- eller komponentnavne | Oxolinic acid ethyl ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H15 N O5 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Ethyl 5-Ethyl-8-oxo-5,8-dihydro-1,3-dioxolo[4,5-g]quinoline-7-carboxylate (Oxolinic Acid Ethyl Ester), Oxolinic Acid Imp. B (EP), 5-ethyl-5,8-dihydro-8-oxo-1,3-Dioxolo[4,5-g]quinoline-7-carboxylic acid ethyl ester, Ethyl oxolinate, Oxolinic Acid Impurity B |
| InChI formel | InChI=1S/C15H15NO5/c1-3-16-7-10(15(18)19-4-2)14(17)9-5-12-13(6-11(9)16)21-8-20-12/h5-7H,3-4,8H2,1-2H3 |
| SMIL | CCOC(=O)C1=CN(CC)c2cc3OCOc3cc2C1=O |
| IUPAC navn | ethyl 5-ethyl-8-oxo-[1,3]dioxolo[4,5-g]quinoline-7-carboxylate |
| Vis mere |