Få mere at vide
Ethyl-5-(4-methoxy-d3-2,3,6-trimethylphenyl)-3-methyl-2,4-pentadienoate(cis/trans Mixture), TRC
Artikelnummer. 30372871
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30372871 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372871 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9250525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl-5-(4-methoxy-d3-2,3,6-trimethylphenyl)-3-methyl-2,4-pentadienoate(cis/trans Mixture)
CAS 1185247-60-2
Molekylær formel C18H21D3O3
Mængde 5 mg
Synonym 5-(4-Methoxy-d3-2,3,6-trimethylphenyl)-3-methyl-2,4-pentadienoic Acid Ethyl Ester
InChI formel InChI=1S/C18H24O3/c1-7-21-18(19)10-12(2)8-9-16-13(3)11-17(20-6)15(5)14(16)4/h8-11H,7H2,1-6H3/b9-8+,12-10+/i6D3
SMIL [2H]C([2H])([2H])Oc1cc(C)c(\C=C\C(=C\C(=O)OCC)\C)c(C)c1C
Molekylvægt (g/mol) 291.4