Få mere at vide
Ethyl 5-(1-Methyl-5-nitro-benzimidazol-2-yl)pentanoate, Mikromol™
Artikelnummer. 30460293
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460293 4 x 25 mg
30051173 100 mg
30049119 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460293 Leverandør Mikromol Leverandørnr. MM0154.5300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 5-(1-Methyl-5-nitro-benzimidazol-2-yl)pentanoate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 5-(1-Methyl-5-nitro-benzimidazol-2-yl)pentanoate
CAS 3543-76-8
Analyt- eller komponentnavne Ethyl 5-(1-Methyl-5-nitro-benzimidazol-2-yl)pentanoate
Forsendelsestilstand Room Temperature
Molekylær formel C15 H19 N3 O4
Certificeringer/compliance ISO 9001
Synonym 1H-Benzimidazole-2-pentanoic acid, 1-methyl-5-nitro-, ethyl ester, 2-Benzimidazolevaleric acid, 1-methyl-5-nitro-, ethyl ester (7CI,8CI)
InChI formel InChI=1S/C15H19N3O4/c1-3-22-15(19)7-5-4-6-14-16-12-10-11(18(20)21)8-9-13(12)17(14)2/h8-10H,3-7H2,1-2H3
Løsningstype Neat
SMIL CCOC(=O)CCCCc1nc2cc(ccc2n1C)[N+](=O)[O-]
IUPAC navn ethyl 5-(1-methyl-5-nitrobenzimidazol-2-yl)pentanoate
Molekylvægt (g/mol) 305.33
Mængde 4 x 25 mg
Formel vægt 305.1376 g/mol