Learn More
Ethyl 4-hydroxycyclohexanecarboxylate, 97%, BLD Pharm™
Product Code. 30804275
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30804275 25 g 25g
30788141 100 g 100g
30802584 1 g 1g
30785862 5 g 5g
30794011 500 g 500g
30787516 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30804275 Supplier BLD Pharm Supplier No. BD12890925g
 Restricted Item
Forventet forsendelse: 25-11-2026
Specifications
CAS 17159-80-7
Molecular Formula C9H16O3
MDL Number MFCD00192156
Quantity 25 g
SMILES O=C(C1CCC(O)CC1)OCC
Molecular Weight (g/mol) 172.22
Percent Purity 97%
Chemical Name or Material Ethyl 4-hydroxycyclohexanecarboxylate