Få mere at vide
Ethyl 4-(Dipropylsulfamoyl)benzoate (Probenecid Ethyl Ester), Mikromol™
Artikelnummer. 30051742
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051742 25 mg
17922806 100 mg
30461491 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051742 Leverandør Mikromol Leverandørnr. MM0657.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 4-(Dipropylsulfamoyl)benzoate (Probenecid Ethyl Ester)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 4-(Dipropylsulfamoyl)benzoate (Probenecid Ethyl Ester)
CAS 70190-76-0
Analyt- eller komponentnavne Ethyl 4-(Dipropylsulfamoyl)benzoate (Probenecid Ethyl Ester)
Forsendelsestilstand Room Temperature
Molekylær formel C15 H23 N O4 S
Certificeringer/compliance ISO 17025
Synonym Probenecid Imp. D (EP), 4-[(Dipropylamino)sulfonyl]benzoic acid ethyl ester, Probenecid Impurity D
InChI formel InChI=1S/C15H23NO4S/c1-4-11-16(12-5-2)21(18,19)14-9-7-13(8-10-14)15(17)20-6-3/h7-10H,4-6,11-12H2,1-3H3
SMIL CCCN(CCC)S(=O)(=O)c1ccc(cc1)C(=O)OCC
IUPAC navn ethyl 4-(dipropylsulfamoyl)benzoate
Molekylvægt (g/mol) 313.41
Mængde 25 mg
Formel vægt 313.1348 g/mol