missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloroquinoline-3-carboxylate, 96%, Thermo Scientific™

Catalog Number 11499260
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
13720-94-0
C12H10ClNO2
235.667
MFCD00173407
DWXQUAHMZWZXHP-UHFFFAOYSA-N
ethyl 4-chloro-3-quinolinecarboxylate, 4-chloroquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-quinoline-3-carboxylate, 3-quinolinecarboxylic acid, 4-chloro-, ethyl ester, pubchem19592, acmc-20ai5d, ethylchloroquinolinecarboxylate, ethyl 4-chloroquinolin-3-carboxylate, ethyl 4-chloro-3-quinoline carboxylate, ethyl 4-chloranylquinoline-3-carboxylate
268963
ethyl 4-chloroquinoline-3-carboxylate
CCOC(=O)C1=C(C2=CC=CC=C2N=C1)Cl
This item is not returnable. View return policy
13720-94-0
C12H10ClNO2
235.667
MFCD00173407
DWXQUAHMZWZXHP-UHFFFAOYSA-N
ethyl 4-chloro-3-quinolinecarboxylate, 4-chloroquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-quinoline-3-carboxylate, 3-quinolinecarboxylic acid, 4-chloro-, ethyl ester, pubchem19592, acmc-20ai5d, ethylchloroquinolinecarboxylate, ethyl 4-chloroquinolin-3-carboxylate, ethyl 4-chloro-3-quinoline carboxylate, ethyl 4-chloranylquinoline-3-carboxylate
268963
ethyl 4-chloroquinoline-3-carboxylate
CCOC(=O)C1=C(C2=CC=CC=C2N=C1)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 4-chloroquinoline-3-carboxylate is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 235.67
Percent Purity 96%
Chemical Name or Material Ethyl 4-chloroquinoline-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'