Få mere at vide
Ethyl 4-Aminopentanoate Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 4-Aminopentanoate Hydrochloride |
| CAS | 119396-97-3 |
| Molekylær formel | C7H15NO2 . HCl |
| Mængde | 10 mg |
| Synonym | Pentanoic acid, 4-amino-, ethyl ester, hydrochloride (1:1) (ACI), Pentanoic acid, 4-amino-, ethyl ester, hydrochloride (9CI), 4-Aminopentanoic acid ethyl ester hydrochloride, 4-Aminovaleric acid ethyl ester hydrochloride, Ethyl 4-aminovalerate hydrochloride, Ethyl 4-aminopentanoate hydrochloride |
| InChI formel | InChI=1S/C7H15NO2.ClH/c1-3-10-7(9)5-4-6(2)8;/h6H,3-5,8H2,1-2H3;1H |
| SMIL | Cl.CCOC(=O)CCC(C)N |
| IUPAC navn | ethyl 4-aminopentanoate;hydrochloride |
| Molekylvægt (g/mol) | 181.66 |
| Formel vægt | 181.087 |