Få mere at vide
Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidine-5-carboxylate-d6, TRC
Artikelnummer. 30371795
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30371795 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371795 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91791250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidine-5-carboxylate-d6
CAS 1189696-04-5
Molekylær formel C18H16D6FN3O4S
Mængde 50 mg
Synonym 4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinecarboxylic Acid Ethyl Ester
InChI formel InChI=1S/C18H22FN3O4S/c1-6-26-17(23)14-15(11(2)3)20-18(22(4)27(5,24)25)21-16(14)12-7-9-13(19)10-8-12/h7-11H,6H2,1-5H3/i4D3,5D3
SMIL [2H]C([2H])([2H])N(c1nc(C(C)C)c(C(=O)OCC)c(n1)c2ccc(F)cc2)S(=O)(=O)C([2H])([2H])[2H]
IUPAC navn ethyl 4-(4-fluorophenyl)-6-propan-2-yl-2-[trideuteriomethyl(trideuteriomethylsulfonyl)amino]pyrimidine-5-carboxylate
Molekylvægt (g/mol) 401.49
Formel vægt 401.1692