Få mere at vide
Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-methylthiopyrimidine-5-carboxylate, TRC
Artikelnummer. 30376745
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376745 25 mg
30371747 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30376745 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91793525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-methylthiopyrimidine-
CAS 147118-27-2
Molekylær formel C17 H19 F N2 O2 S
Mængde 25 mg
Synonym 5-Pyrimidinecarboxylic acid, 4-(4-fluorophenyl)-6-(1-methylethyl)-2-(methylthio)-, ethyl ester, Ethyl 4-(4-fluorophenyl)-6-isopropyl-2-methylsulfanyl-pyrimidine-5-carboxylate
InChI formel InChI=1S/C17H19FN2O2S/c1-5-22-16(21)13-14(10(2)3)19-17(23-4)20-15(13)11-6-8-12(18)9-7-11/h6-10H,5H2,1-4H3
SMIL CCOC(=O)c1c(nc(SC)nc1c2ccc(F)cc2)C(C)C
IUPAC navn ethyl 4-(4-fluorophenyl)-2-methylsulfanyl-6-propan-2-ylpyrimidine-5-carboxylate
Molekylvægt (g/mol) 334.41
Formel vægt 334.1151