Få mere at vide
Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate, Mikromol™
Artikelnummer. 30050109
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050109 25 mg
30050392 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050109 Leverandør Mikromol Leverandørnr. MM0936.090025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate
CAS 147118-30-7
Analyt- eller komponentnavne Ethyl 4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C18 H22 F N3 O4 S
Certificeringer/compliance ISO 17025
Synonym 5-Pyrimidinecarboxylic acid, 4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-, ethyl ester, Ethyl 4-(4-fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidine-5-carboxylate
InChI formel InChI=1S/C18H22FN3O4S/c1-6-26-17(23)14-15(11(2)3)20-18(22(4)27(5,24)25)21-16(14)12-7-9-13(19)10-8-12/h7-11H,6H2,1-5H3
SMIL CCOC(=O)c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C
IUPAC navn ethyl 4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidine-5-carboxylate
Molekylvægt (g/mol) 395.45
Mængde 25 mg
Formel vægt 395.1315 g/mol